siRNA Details
| Gene Symbol: Eldr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK141825.1 | - | 2 | 3027 | ||
| AK163834.1 | - | 1 | 1676 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eldr| Gene Symbol: Eldr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK141825.1 | - | 2 | 3027 | ||
| AK163834.1 | - | 1 | 1676 | |||