siRNA Details
| Gene Symbol: 1700020I14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015473.1 | 3 | 854 | |||
| NR_027832.1 | 3 | 685 | ||||
| GenBank mRNA | AK006166.1 | - | 3 | 824 | ||
| AK009929.1 | - | 3 | 661 | |||
| AK011393.1 | - | 3 | 818 | |||
| AK011848.1 | - | 3 | 820 | |||
| AK017100.1 | - | 3 | 686 | |||
| AK029148.1 | - | 820 | ||||
| AK030275.1 | - | 817 | ||||
| AK147270.1 | - | 831 | ||||
| AK170765.1 | - | 3 | 824 | |||
| BC026913.1 | - | 3 | 651 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ964656.1 | - | 3 | 801 | Chr. 2 - 119594362 - 119600241 [+] on Build GRCm38 | |

1700020I14Rik
Genomic Map