siRNA Details
| Gene Symbol: 1700020I14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015473.1 | 3 | 1047 | |||
| NR_027832.1 | 3 | 878 | ||||
| GenBank mRNA | AK006166.1 | - | 3 | 1017 | ||
| AK009929.1 | - | 3 | 854 | |||
| AK011393.1 | - | 3 | 1011 | |||
| AK017100.1 | - | 3 | 879 | |||
| AK029148.1 | - | 1013 | ||||
| AK147270.1 | - | 1024 | ||||
| BC026913.1 | - | 3 | 844 | |||

1700020I14Rik
Genomic Map