siRNA Details
| Gene Symbol: Lrrc57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027510.1 | 1, 2 | 233 | |||
| GenBank mRNA | AK134971.1 | - | 1, 2 | 84 | ||
| BY081522.1 | - | 1, 2 | 234 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc57| Gene Symbol: Lrrc57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027510.1 | 1, 2 | 233 | |||
| GenBank mRNA | AK134971.1 | - | 1, 2 | 84 | ||
| BY081522.1 | - | 1, 2 | 234 | |||