siRNA Details
| Gene Symbol: Vax2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_002873.2 | 2 | 1909 | |||
| GenBank mRNA | AY589787.1 | - | LIT3407 Vax2OS |
2 | 1913 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vax2os| Gene Symbol: Vax2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_002873.2 | 2 | 1909 | |||
| GenBank mRNA | AY589787.1 | - | LIT3407 Vax2OS |
2 | 1913 | |