siRNA Details
| Gene Symbol: Gm8234 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_004433.3 | 1 | 744 | |||
| GenBank mRNA | AA241209.1 | - | 1 | 323 | ||
| AI508387.1 | - | 1 | 323 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm8234| Gene Symbol: Gm8234 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_004433.3 | 1 | 744 | |||
| GenBank mRNA | AA241209.1 | - | 1 | 323 | ||
| AI508387.1 | - | 1 | 323 | |||