siRNA Details
| Gene Symbol: D6Ertd474e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027803.1 | 4 | 1779 | |||
| GenBank mRNA | AK139687.1 | - | 4 | 1775 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D6Ertd474e| Gene Symbol: D6Ertd474e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027803.1 | 4 | 1779 | |||
| GenBank mRNA | AK139687.1 | - | 4 | 1775 | ||