siRNA Details
| Gene Symbol: Peg13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_002864.1 | 1 | 4572 | |||
| GenBank mRNA | AY151252.1 | - | LIT1829 Peg13 |
1 | 4572 | |
| AY151253.1 | - | LIT1828 Peg13 |
1 | 4246 | ||

Peg13
Genomic Map