siRNA Details
| Gene Symbol: 5830432E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015548.1 | 3 | 1084 | |||
| GenBank mRNA | AK017962.1 | - | 3 | 1085 | ||
| AK020025.1 | - | 3 | 1082 | |||
| BY548245.1 | - | 1 | 20 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5830432E09Rik| Gene Symbol: 5830432E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015548.1 | 3 | 1084 | |||
| GenBank mRNA | AK017962.1 | - | 3 | 1085 | ||
| AK020025.1 | - | 3 | 1082 | |||
| BY548245.1 | - | 1 | 20 | |||