siRNA Details
| Gene Symbol: Fam172a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028109.1 | 3, 4 | 371 | |||
| GenBank mRNA | AK165273.1 | - | 3, 4 | 345 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam172a| Gene Symbol: Fam172a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028109.1 | 3, 4 | 371 | |||
| GenBank mRNA | AK165273.1 | - | 3, 4 | 345 | ||