siRNA Details
| Gene Symbol: 6430411K18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002848.3 | 1 | 449 | |||
| GenBank mRNA | AK018276.1 | - | 1 | 463 | ||
| EU434921.1 | - | 1433 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6430411K18Rik| Gene Symbol: 6430411K18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002848.3 | 1 | 449 | |||
| GenBank mRNA | AK018276.1 | - | 1 | 463 | ||
| EU434921.1 | - | 1433 | ||||