siRNA Details
| Gene Symbol: 6430411K18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002848.3 | 1 | 943 | |||
| GenBank mRNA | AK018276.1 | - | 1 | 957 | ||
| EU434921.1 | - | 1927 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB590426.1 | - | 1 | 304 | Chr. 12 - 109592176 - 109592882 [+] on Build GRCm38 | |

6430411K18Rik
Genomic Map