siRNA Details
| Gene Symbol: 1700096K18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027388.1 | 1 | 520 | |||
| GenBank mRNA | AK007094.1 | - | 1 | 526 | ||
| AK047471.1 | - | 1 | 521 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700096K18Rik| Gene Symbol: 1700096K18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027388.1 | 1 | 520 | |||
| GenBank mRNA | AK007094.1 | - | 1 | 526 | ||
| AK047471.1 | - | 1 | 521 | |||