siRNA Details
Gene Symbol: 1700096K18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027388.1 | 1 | 557 | |||
GenBank mRNA | AK047471.1 | - | 1 | 558 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700096K18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027388.1 | 1 | 557 | |||
GenBank mRNA | AK047471.1 | - | 1 | 558 |