siRNA Details
| Gene Symbol: 1700096K18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027388.1 | 1 | 997 | |||
| GenBank mRNA | AK047471.1 | - | 1 | 998 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700096K18Rik| Gene Symbol: 1700096K18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027388.1 | 1 | 997 | |||
| GenBank mRNA | AK047471.1 | - | 1 | 998 | ||