siRNA Details
Gene Symbol: Fbxo34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027414.1 | 2, 3 | 304 | |||
GenBank mRNA | AK013711.1 | - | 2, 3 | 241 | ||
AK171630.1 | - | 3, 4 | 308 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fbxo34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027414.1 | 2, 3 | 304 | |||
GenBank mRNA | AK013711.1 | - | 2, 3 | 241 | ||
AK171630.1 | - | 3, 4 | 308 |