siRNA Details
| Gene Symbol: Fbxo34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027414.1 | 4 | 627 | |||
| GenBank mRNA | AK013711.1 | - | 4 | 564 | ||
| BC048149.1 | - | 2 | 236 | 5706344 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo34| Gene Symbol: Fbxo34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027414.1 | 4 | 627 | |||
| GenBank mRNA | AK013711.1 | - | 4 | 564 | ||
| BC048149.1 | - | 2 | 236 | 5706344 | ||