siRNA Details
Gene Symbol: Fbxo34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027414.1 | 4 | 573 | |||
GenBank mRNA | AK013711.1 | - | 4 | 510 | ||
BC048149.1 | - | 2 | 182 | 5706344 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fbxo34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027414.1 | 4 | 573 | |||
GenBank mRNA | AK013711.1 | - | 4 | 510 | ||
BC048149.1 | - | 2 | 182 | 5706344 |