siRNA Details
| Gene Symbol: Zfp326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030777.1 | 3 | 230 | |||
| GenBank mRNA | AK032801.1 | - | 3 | 231 | ||
| BC037055.1 | - | 3 | 235 | 4948371 | ||
| BU503985.1 | - | 2 | 78 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp326| Gene Symbol: Zfp326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030777.1 | 3 | 230 | |||
| GenBank mRNA | AK032801.1 | - | 3 | 231 | ||
| BC037055.1 | - | 3 | 235 | 4948371 | ||
| BU503985.1 | - | 2 | 78 | |||