siRNA Details
Gene Symbol: 2810454H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029441.1 | 1 | 578 | |||
GenBank mRNA | AK028630.1 | - | 1 | 578 | ||
AK051179.1 | - | 1 | 78 | |||
AK080385.1 | - | 1 | 145 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2810454H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029441.1 | 1 | 578 | |||
GenBank mRNA | AK028630.1 | - | 1 | 578 | ||
AK051179.1 | - | 1 | 78 | |||
AK080385.1 | - | 1 | 145 |