siRNA Details
| Gene Symbol: 2810454H06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029441.1 | 1 | 2650 | |||
| GenBank mRNA | AK028630.1 | - | 1 | 2650 | ||
| AK051179.1 | - | 1 | 2150 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810454H06Rik| Gene Symbol: 2810454H06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029441.1 | 1 | 2650 | |||
| GenBank mRNA | AK028630.1 | - | 1 | 2650 | ||
| AK051179.1 | - | 1 | 2150 | |||