siRNA Details
| Gene Symbol: 2810454H06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029441.1 | 1 | 756 | |||
| GenBank mRNA | AK028630.1 | - | 1 | 756 | ||
| AK051179.1 | - | 1 | 256 | |||
| AK080385.1 | - | 1 | 323 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB639297.1 | - | 1 | 320 | Chr. 6 - 134898397 - 134899084 [+] on Build GRCm38 | |

2810454H06Rik
Genomic Map