siRNA Details
| Gene Symbol: C86187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015609.1 | 2 | 1133 | |||
| GenBank mRNA | AK135809.1 | - | 2 | 1130 | ||
| AK135845.1 | - | 4 | 1496 | |||
| AK135966.1 | - | 2 | 1135 | |||
| AK139730.1 | - | 2 | 1135 | |||
| AK163234.1 | - | 2 | 1132 | |||

C86187
Genomic Map