siRNA Details
| Gene Symbol: C86187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015609.1 | 1 | 531 | |||
| GenBank mRNA | AK135809.1 | - | 1 | 528 | ||
| AK135845.1 | - | 1 | 534 | |||
| AK135966.1 | - | 1 | 533 | |||
| AK139730.1 | - | 1 | 532 | |||
| AK162109.1 | - | 1 | 532 | |||
| AK163234.1 | - | 1 | 530 | |||

C86187
Genomic Map