siRNA Details
| Gene Symbol: A530088E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029458.1 | 1 | 566 | |||
| GenBank mRNA | AK041179.1 | - | 1 | 559 | ||
| AK046549.1 | - | 1 | 563 | |||
| AK162668.1 | - | 1 | 548 | |||
| AK169535.1 | - | 1 | 576 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A530088E08Rik| Gene Symbol: A530088E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029458.1 | 1 | 566 | |||
| GenBank mRNA | AK041179.1 | - | 1 | 559 | ||
| AK046549.1 | - | 1 | 563 | |||
| AK162668.1 | - | 1 | 548 | |||
| AK169535.1 | - | 1 | 576 | |||