siRNA Details
| Gene Symbol: 2810429I04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015522.1 | 3 | 145 | |||
| GenBank mRNA | AK013191.1 | - | 153 | |||
| AK031301.1 | - | 131 | ||||
| AK034854.1 | - | 1 | 1709 | |||
| AK043092.1 | - | 3 | 227 | |||
| AK043278.1 | - | 3 | 146 | |||
| AK084238.1 | - | 1 | 1719 | |||
| AK087718.1 | - | 2 | 140 | |||

2810429I04Rik
Genomic Map