siRNA Details
| Gene Symbol: Rreb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033218.1 | 12 | 9279 | |||
| NR_033615.1 | 10 | 5974 | ||||
| GenBank mRNA | AK004146.1 | - | 1 | 1387 | ||
| AK147260.1 | - | 12 | 9280 | |||
| AK147653.1 | - | 12 | 9280 | |||
| AY946044.1 | - | 10 | 7671 | |||

Rreb1
Genomic Map