siRNA Details
| Gene Symbol: Rreb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033218.1 | 12 | 7027 | |||
| NR_033615.1 | 10 | 3722 | ||||
| GenBank mRNA | AK147260.1 | - | 12 | 7028 | ||
| AK147653.1 | - | 12 | 7028 | |||
| AY946044.1 | - | 10 | 5419 | |||
| BC027834.1 | - | 2 | 1282 | |||

Rreb1
Genomic Map