siRNA Details
| Gene Symbol: R74862 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015529.2 | 4 | 1199 | |||
| NR_110354.1 | 4 | 1199 | ||||
| NR_110355.1 | 5 | 1334 | ||||
| GenBank mRNA | AK034864.1 | - | 5 | 1326 | ||
| AK035397.1 | - | 4 | 882 | |||
| AK083267.1 | - | 4 | 931 | |||
| BC005669.1 | - | 4 | 616 | |||
| BC080794.1 | - | 4 | 1199 | |||

R74862
Genomic Map