siRNA Details
| Gene Symbol: R74862 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015529.2 | 4 | 1999 | |||
| NR_110355.1 | 5 | 2134 | ||||
| GenBank mRNA | AK035397.1 | - | 4 | 1682 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

R74862| Gene Symbol: R74862 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015529.2 | 4 | 1999 | |||
| NR_110355.1 | 5 | 2134 | ||||
| GenBank mRNA | AK035397.1 | - | 4 | 1682 | ||