siRNA Details
| Gene Symbol: R74862 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015529.2 | 4 | 945 | |||
| NR_110354.1 | 4 | 945 | ||||
| NR_110355.1 | 5 | 1080 | ||||
| GenBank mRNA | AK034864.1 | - | 5 | 1072 | ||
| AK035397.1 | - | 4 | 628 | |||
| AK083267.1 | - | 4 | 677 | |||
| BC005669.1 | - | 4 | 362 | |||
| BC080794.1 | - | 4 | 945 | |||

R74862
Genomic Map