siRNA Details
| Gene Symbol: Mir218-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029798.1 | 1 | 59 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mir218-1| Gene Symbol: Mir218-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029798.1 | 1 | 59 | |||