siRNA Details
| Gene Symbol: 4632415L05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027985.1 | 1 | 2679 | |||
| GenBank mRNA | BC037267.1 | - | 1 | 2652 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4632415L05Rik| Gene Symbol: 4632415L05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027985.1 | 1 | 2679 | |||
| GenBank mRNA | BC037267.1 | - | 1 | 2652 | ||