siRNA Details
Gene Symbol: 4632415L05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027985.1 | 1 | 3123 | |||
GenBank mRNA | BC037267.1 | - | 1 | 3096 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4632415L05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027985.1 | 1 | 3123 | |||
GenBank mRNA | BC037267.1 | - | 1 | 3096 |