siRNA Details
| Gene Symbol: Gm15760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030670.1 | 1 | 667 | |||
| GenBank mRNA | AK149512.1 | - | 1 | 568 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15760| Gene Symbol: Gm15760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030670.1 | 1 | 667 | |||
| GenBank mRNA | AK149512.1 | - | 1 | 568 | ||