siRNA Details
| Gene Symbol: 4930474M22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027986.1 | 3 | 350 | |||
| GenBank mRNA | AK015576.1 | - | 3 | 353 | ||
| BC089489.1 | - | 3 | 328 | 6741101 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_183107.1 | - | 353 | Chr. - - [-] on Build | ||

4930474M22Rik
Genomic Map