siRNA Details
| Gene Symbol: Snhg20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027059.1 | 1 | 193 | |||
| GenBank mRNA | AK012687.1 | - | 1 | 187 | ||
| AK171830.1 | - | 1 | 175 | |||
| BC037520.1 | - | 1 | 113 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snhg20| Gene Symbol: Snhg20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027059.1 | 1 | 193 | |||
| GenBank mRNA | AK012687.1 | - | 1 | 187 | ||
| AK171830.1 | - | 1 | 175 | |||
| BC037520.1 | - | 1 | 113 | |||