siRNA Details
| Gene Symbol: Dctd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027759.1 | 5 | 1169 | |||
| GenBank mRNA | AK171894.1 | - | 5 | 966 | ||
| BQ572929.1 | - | 3 | 643 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dctd| Gene Symbol: Dctd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027759.1 | 5 | 1169 | |||
| GenBank mRNA | AK171894.1 | - | 5 | 966 | ||
| BQ572929.1 | - | 3 | 643 | |||