siRNA Details
| Gene Symbol: Pmis2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027848.1 | 2 | 303 | |||
| NR_027849.1 | 2 | 169 | ||||
| GenBank mRNA | AB690425.1 | - | 2 | 225 | ||
| AK006932.1 | - | 2 | 171 | |||
| AK015595.1 | - | 2 | 283 | |||
| BC100424.1 | - | 2 | 289 | 30920315 | ||

Pmis2
Genomic Map