siRNA Details
| Gene Symbol: Pmis2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027848.1 | 2 | 680 | |||
| NR_027849.1 | 2 | 546 | ||||
| GenBank mRNA | AK015595.1 | - | 2 | 660 | ||
| BC100424.1 | - | 2 | 666 | 30920315 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_183108.1 | - | 660 | Chr. - - [-] on Build | ||

Pmis2
Genomic Map