siRNA Details
| Gene Symbol: Pmis2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027848.1 | 1 | 40 | |||
| NR_027849.1 | 1 | 40 | ||||
| GenBank mRNA | AK006932.1 | - | 1 | 42 | ||
| AK015595.1 | - | 1 | 20 | |||
| BC100424.1 | - | 1 | 26 | 30920315 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pmis2| Gene Symbol: Pmis2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027848.1 | 1 | 40 | |||
| NR_027849.1 | 1 | 40 | ||||
| GenBank mRNA | AK006932.1 | - | 1 | 42 | ||
| AK015595.1 | - | 1 | 20 | |||
| BC100424.1 | - | 1 | 26 | 30920315 | ||