siRNA Details
| Gene Symbol: Snhg12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029468.1 | 5 | 561 | |||
| GenBank mRNA | AK009175.1 | - | 4 | 894 | ||
| CB201672.1 | - | 5 | 546 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA060788.1 | - | 5 | 426 | Chr. 4 - 132308813 - 132311023 [+] on Build GRCm38 | |

Snhg12
Genomic Map