siRNA Details
| Gene Symbol: 4933400A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003635.1 | 3 | 909 | |||
| GenBank mRNA | AK015979.1 | - | 2 | 776 | ||
| AK016585.1 | - | 3 | 910 | |||
| BC107240.1 | - | 3 | 827 | |||
| BC107241.1 | - | 3 | 831 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933400A11Rik| Gene Symbol: 4933400A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003635.1 | 3 | 909 | |||
| GenBank mRNA | AK015979.1 | - | 2 | 776 | ||
| AK016585.1 | - | 3 | 910 | |||
| BC107240.1 | - | 3 | 827 | |||
| BC107241.1 | - | 3 | 831 | |||