siRNA Details
| Gene Symbol: 4933404O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015555.1 | 4 | 1339 | |||
| GenBank mRNA | AK085506.1 | - | 4 | 1342 | ||
| AK169739.1 | - | 4 | 1422 | |||
| CV784581.1 | - | 1 | 261 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933404O12Rik| Gene Symbol: 4933404O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015555.1 | 4 | 1339 | |||
| GenBank mRNA | AK085506.1 | - | 4 | 1342 | ||
| AK169739.1 | - | 4 | 1422 | |||
| CV784581.1 | - | 1 | 261 | |||