siRNA Details
Gene Symbol: 4933404O12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015555.1 | 4 | 1215 | |||
GenBank mRNA | AK085506.1 | - | 4 | 1218 | ||
AK169739.1 | - | 4 | 1297 | |||
CV784581.1 | - | 1 | 137 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU514324.1 | - | 2 | 696 | Chr. 5 - 136925850 - 136934624 [+] on Build GRCm38 |
