siRNA Details
| Gene Symbol: Gm5129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028426.1 | 1 | 454 | |||
| GenBank mRNA | AK076721.1 | - | 1 | 457 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5129| Gene Symbol: Gm5129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028426.1 | 1 | 454 | |||
| GenBank mRNA | AK076721.1 | - | 1 | 457 | ||