siRNA Details
| Gene Symbol: Gm5129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028426.1 | 1 | 494 | |||
| GenBank mRNA | AK076721.1 | - | 1 | 497 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB615089.1 | - | 1 | 487 | Chr. 5 - 29735353 - 29735929 [-] on Build GRCm38 | |

Gm5129
Genomic Map