siRNA Details
| Gene Symbol: E030024N20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033228.1 | 1 | 3654 | |||
| GenBank mRNA | AK142495.1 | - | 1 | 3654 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E030024N20Rik| Gene Symbol: E030024N20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033228.1 | 1 | 3654 | |||
| GenBank mRNA | AK142495.1 | - | 1 | 3654 | ||