siRNA Details
| Gene Symbol: Rcbtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033185.1 | 4, 5 | 658 | |||
| GenBank mRNA | AK147784.1 | - | 4, 5 | 279 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rcbtb2| Gene Symbol: Rcbtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033185.1 | 4, 5 | 658 | |||
| GenBank mRNA | AK147784.1 | - | 4, 5 | 279 | ||