siRNA Details
| Gene Symbol: Rcbtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033185.1 | 1 | 18 | |||
| GenBank mRNA | AK013119.1 | - | 1 | 95 | ||
| BB854398.1 | - | 1 | 18 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rcbtb2| Gene Symbol: Rcbtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033185.1 | 1 | 18 | |||
| GenBank mRNA | AK013119.1 | - | 1 | 95 | ||
| BB854398.1 | - | 1 | 18 | |||